CAS RN 2648-71-7
IUPAC Name:
3-bromo-3-methylbutan-2-one
Synonyms:
3-bromo-3-methylbutan-2-one
Molecular Weight (g/mol):
165.03
Molecular Formula:
C5H9BrO
InChi Key:
XHCWDDGZFBAXRT-UHFFFAOYSA-N
SMILES:
CC(=O)C(C)(C)Br
1
–
3
of
3
results
1
3-Bromo-3-methyl-2-butanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Bromo-3-methyl-2-butanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Bromo-3-methyl-2-butanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.