accessibility menu, dialog, popup

UserName

CAS RN 2650-53-5

IUPAC Name: hexyltrimethylazanium bromide
Synonyms: hexyltrimethylazanium bromide
Molecular Weight (g/mol): 224.19
Molecular Formula: C9H22BrN
InChi Key: JYVPKRHOTGQJSE-UHFFFAOYSA-M
SMILES: [Br-].CCCCCC[N+](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results