CAS RN 265121-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fosaprepitant Dibenzyl Ester, TRC
CAS: 265121-01-5 Molecular Formula: C37H34F7N4O6P Molecular Weight (g/mol): 794.65 Synonym: [3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(Trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-2,5-dihydro-5-oxo-1H-1,2,4-triazol-1-yl]-phosphonic Acid bis(Phenylmethyl) Ester,Dibenzyl Fosaprepitant; IUPAC Name: dibenzyl (3-(((2R,3S)-2-((R)-1-(3,5-bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-5-oxo-2,5-dihydro-1H-1,2,4-triazol-1-yl)phosphonate SMILES: O=C1N=C(CN2[C@@H](C3=CC=C(F)C=C3)[C@@H](O[C@@H](C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)C)OCC2)NN1P(OCC5=CC=CC=C5)(OCC6=CC=CC=C6)=O