CAS RN 26522-85-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Malonic Acid Disodium Salt Monohydrate, TRC
CAS: 26522-85-0 Molecular Formula: C3H2Na2O4 . H2O Molecular Weight (g/mol): 166.04 IUPAC Name: disodium;propanedioate;hydrate SMILES: O.[Na+].[Na+].[O-]C(=O)CC(=O)[O-]