CAS RN 2653-16-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-N-methyl-N-(4-nitrophenyl)acetamide, TRC
CAS: 2653-16-9 Molecular Formula: C9H9ClN2O3 Molecular Weight (g/mol): 228.63 Synonym: α-Chloro-N-methyl-p-nitroacetanilide,2-Chloro-N-methyl-4'-nitro-acetanilide,2-Chloro-N-methyl-N-(4-nitrophenyl)-acetamide IUPAC Name: 2-chloro-N-methyl-N-(4-nitrophenyl)acetamide SMILES: CN(C(=O)CCl)c1ccc(cc1)[N+](=O)[O-]