CAS RN 26530-20-1
IUPAC Name:
2-octyl-2,3-dihydro-1,2-thiazol-3-one
Synonyms:
octylisothiazolinone
Molecular Weight (g/mol):
213.34
Molecular Formula:
C11H19NOS
InChi Key:
JPMIIZHYYWMHDT-UHFFFAOYSA-N
SMILES:
CCCCCCCCN1SC=CC1=O
1
–
1
of
1
results
1
Octhilinone, TRC
CAS: 26530-20-1 Molecular Formula: C11 H19 N O S Molecular Weight (g/mol): 213.34 Synonym: 2-Octyl-2H-isothiazol-3-one,OIT IUPAC Name: 2-octyl-1,2-thiazol-3-one SMILES: CCCCCCCCN1SC=CC1=O