CAS RN 26539-01-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(3,5-Dihydroxyphenyl)-1-propanoic Acid, TRC
CAS: 26539-01-5 Molecular Formula: C9H10O4 Molecular Weight (g/mol): 182.17 Synonym: (8CI) 3,5-Dihydroxyhydrocinnamic Acid,3,5-Dihydroxybenzenepropanoic Acid,3,5-Dihydroxyhydrocinnamic Acid; SMILES: OC(=O)CCc1cc(O)cc(O)c1