accessibility menu, dialog, popup

UserName

CH3OBr

CAS RN 2655-84-7

IUPAC Name: 1-bromo-3-ethoxybenzene
Synonyms: 1-bromo-3-ethoxybenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: LQBMPJSTUHWGDE-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results