CAS RN 2657-87-6
IUPAC Name:
4-(3-aminophenoxy)aniline
Synonyms:
4-(3-aminophenoxy)aniline
Molecular Weight (g/mol):
200.24
Molecular Formula:
C12H12N2O
InChi Key:
ZBMISJGHVWNWTE-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OC2=CC=CC(N)=C2)C=C1
1
–
1
of
1
results
1
3,4'-Diaminodiphenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.