CAS RN 2658-74-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
6beta-Chloro-17-acetoxy Progesterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6b-Chloro-17-acetoxy Progesterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6b-Chloro-17-acetoxy Progesterone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
6Alpha-Chloro-17-acetoxy Progesterone, TRC
Molecular Formula: C23 H31 Cl O4