accessibility menu, dialog, popup

UserName

NH3CH3CCH3CH2Cl

CAS RN 26616-35-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 213.75
Molecular Formula: C12H20ClN
InChi Key:
SMILES: [Cl-].*-C=C.C[N+](C)(C)C-*.C1=CC=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)