CAS RN 266359-93-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Reparixin L-lysine salt, MedChemExpress
MedChemExpress Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation.
2
Tocris™ Reparixin L-lysine salt
Potent CXCR1 and CXCR2 allosteric antagonist