CAS RN 26638-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
rac 3,11-Dichloro-6,11-dihydro-6-methyl-dibenzo[c,f][1,2]thiazepine 5,5-Dioxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
rac 3,11-Dichloro-6,11-dihydro-6-methyl-dibenzo[c,f][1,2]thiazepine 5,5-Dioxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.