CAS RN 2670-67-9
IUPAC Name:
1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium hydrobromide
Synonyms:
pinacyanol(1+) hydrobromide
Molecular Weight (g/mol):
434.4
Molecular Formula:
C25H26BrN2
InChi Key:
COPIHGSSCONEQG-UHFFFAOYSA-N
SMILES:
Br.CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12