accessibility menu, dialog, popup

UserName

CAS RN 267221-88-5

BOOH3CCH3H3CH3CN

CAS RN 267221-88-5

IUPAC Name: N,N-diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: N,N-diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 371.29
Molecular Formula: C24H26BNO2
InChi Key: VKSWIFGDKIEVFZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results