CAS RN 2675-89-0
CAS RN 2675-89-0
IUPAC Name:
2-chloro-N,N-dimethylacetamide
Synonyms:
2-chloro-N,N-dimethylacetamide
Molecular Weight (g/mol):
121.56
Molecular Formula:
C4H8ClNO
InChi Key:
XBPPLECAZBTMMK-UHFFFAOYSA-N
SMILES:
CN(C)C(=O)CCl
1
–
2
of
2
results
1
2-Chloro-N,N-dimethylacetamide, 97%
CAS: 2675-89-0 Molecular Formula: C4H8ClNO MDL Number: MFCD00039360 InChI Key: XBPPLECAZBTMMK-UHFFFAOYSA-N PubChem CID: 75886 IUPAC Name: 2-chloro-N,N-dimethylacetamide SMILES: CN(C)C(=O)CCl
2
2-chloro-N,N-dimethylacetamide, 97%
CAS: 2675-89-0 Molecular Formula: C4H8ClNO Molecular Weight (g/mol): 121.57 InChI Key: XBPPLECAZBTMMK-UHFFFAOYSA-N Synonym: n,n-dimethyl-2-chloroacetamide,n,n-dimethylchloroacetamide,acetamide, 2-chloro-n,n-dimethyl,2-chloro-n, n-dimethylacetamide,acmc-209gsx,2-chlorodimethylacetamide,2-chloro-dimethylacetamide,2-chlorodimethyl acetamide,chloro-n,n-dimethylacetamide,n,n-dimethyl chloroacetamide PubChem CID: 75886 IUPAC Name: 2-chloro-N,N-dimethylacetamide SMILES: CN(C)C(=O)CCl