CAS RN 26763-19-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Macherey-Nagel™ Lyophilized Carrier RNA
Lyophilized,Carrier RNA to be used with NucleoSpin RNA XS or RNA Virus kits.