accessibility menu, dialog, popup

UserName

CH3NOCl

CAS RN 26772-93-0

IUPAC Name: N-(4-chlorophenyl)-N-methylformamide
Synonyms: N-(4-chlorophenyl)-N-methylformamide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: XWQPXHSGRKRZPV-UHFFFAOYSA-N
SMILES: CN(C=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)