CAS RN 2679-01-8
IUPAC Name:
3,7-bis(dimethylamino)-4-nitro-5λ⁴-phenothiazin-5-ylium chloride
Synonyms:
methylene green
Molecular Weight (g/mol):
364.85
Molecular Formula:
C16H17ClN4O2S
InChi Key:
YYGBVRCTHASBKD-UHFFFAOYSA-M
SMILES:
[Cl-].CN(C)C1=CC=C2N=C3C=CC(N(C)C)=C(C3=[S+]C2=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Methylene green, MP Biomedicals™
CAS: 2679-01-8 Molecular Formula: C16H17ClN4O2S Molecular Weight (g/mol): 364.85 InChI Key: YYGBVRCTHASBKD-UHFFFAOYSA-M Synonym: C.I. 52020,3, 7-Bis(Dimethylamino)-4-Nitro-Phenothiazin-5-Ium Chloride