accessibility menu, dialog, popup

UserName

O2H2H2H2H2H2H2H2H2H2H

CAS RN 2679-89-2

IUPAC Name: 1-[(1,1,2,2,2-²H₅)ethoxy](²H₅)ethane
Synonyms: 1-[(1,1,2,2,2-²H₅)ethoxy](²H₅)ethane
Molecular Weight (g/mol): 84.18
Molecular Formula: C4H10O
InChi Key: RTZKZFJDLAIYFH-MWUKXHIBSA-N
SMILES: [2H]C([2H])([2H])C([2H])([2H])OC([2H])([2H])C([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results