accessibility menu, dialog, popup

UserName

NNH2

CAS RN 26807-73-8

IUPAC Name: 1-benzyl-1H-indol-5-amine
Synonyms: 1-benzylindol-5-amine
Molecular Weight (g/mol): 222.29
Molecular Formula: C15H14N2
InChi Key: UYDNPZLYDODKKA-UHFFFAOYSA-N
SMILES: NC1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results