CAS RN 2681-94-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-2-Bromobutyric Acid, TRC
CAS: 2681-94-9 Molecular Formula: C4 H7 Br O2 Molecular Weight (g/mol): 167 Synonym: Butanoic acid, 2-bromo-, (2R)- (9CI, ACI),(2R)-2-Bromobutanoic acid (ACI),Butanoic acid, 2-bromo-, (R)- (ZCI),Butyric acid, 2-bromo-, D- (8CI),(2R)-2-Bromobutanoic acid,(R)-2-Bromobutanoic acid,(R)-2-Bromobutyric acid,(R)-2-Bromobutyric acid,(R)-α-Bromobutyric acid,D-2-Bromobutanoic acid,D-2-Bromobutyric acid IUPAC Name: (2R)-2-bromobutanoic acid SMILES: CC[C@@H](Br)C(=O)O