accessibility menu, dialog, popup

UserName

HClCH3NSO

CAS RN 2682-20-4

IUPAC Name: hydrogen 2-methyl-2,3-dihydro-1,2-thiazol-3-one chloride
Synonyms: methylisothiazolinone
Molecular Weight (g/mol): 151.61
Molecular Formula: C4H6ClNOS
InChi Key: SJXPQSRCFCPWQQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1SC=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results