CAS RN 2687960-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sodium D-Gluconate-13C6, TRC
CAS: 2687960-00-3 Molecular Formula: 13C6H11NaO7 Molecular Weight (g/mol): 224.09 Synonym: D-Gluconic-13C6 Acid Sodium Salt; IUPAC Name: sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate-1,2,3,4,5,6-13C6 SMILES: O[13C@H]([13C@@H](O)[13C](O[Na])=O)[13C@H](O)[13C@H](O)[13CH2]O