accessibility menu, dialog, popup

UserName

FFFFFFN

CAS RN 27126-93-8

IUPAC Name: 3,5-bis(trifluoromethyl)benzonitrile
Synonyms: 3,5-bis(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 239.12
Molecular Formula: C9H3F6N
InChi Key: CZKHHAOIHXHOSR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results