accessibility menu, dialog, popup

UserName

CAS RN 2719-27-9

ClO

CAS RN 2719-27-9

IUPAC Name: cyclohexanecarbonyl chloride
Synonyms: cyclohexanecarbonyl chloride
Molecular Weight (g/mol): 146.61
Molecular Formula: C7H11ClO
InChi Key: RVOJTCZRIKWHDX-UHFFFAOYSA-N
SMILES: ClC(=O)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)