accessibility menu, dialog, popup

UserName

H2COHO

CAS RN 27326-43-8

IUPAC Name: 2-ethenylbenzoic acid
Synonyms: 2-ethenylbenzoic acid
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: XUDBVJCTLZTSDC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results