accessibility menu, dialog, popup

UserName

CAS RN 279263-10-4

CH3OFBOHHO

CAS RN 279263-10-4

IUPAC Name: (4-ethoxy-3-fluorophenyl)boronic acid
Synonyms: 4-ethoxy-3-fluorophenylboronic acid
Molecular Weight (g/mol): 183.97
Molecular Formula: C8H10BFO3
InChi Key: ZONJMULNQBNBGU-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1F)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results