accessibility menu, dialog, popup

UserName

CAS RN 2803-00-1

NNOFFF

CAS RN 2803-00-1

IUPAC Name: 1-(4-benzylpiperazin-1-yl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(4-benzylpiperazin-1-yl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 272.27
Molecular Formula: C13H15F3N2O
InChi Key: VZIHGMYACKUQQJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)N1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results