CAS RN 280566-45-2
CAS RN 280566-45-2
IUPAC Name:
2-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms:
2-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol):
225.55
Molecular Formula:
C7H3ClF3NO2
InChi Key:
DXRBTBMFFGEVCX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Cl)N=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Chloro-6-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™
CAS: 280566-45-2 Molecular Formula: C7H3ClF3NO2 Molecular Weight (g/mol): 225.551 MDL Number: MFCD01862658 InChI Key: DXRBTBMFFGEVCX-UHFFFAOYSA-N Synonym: 2-chloro-6-trifluoromethyl nicotinic acid,2-chloro-6-trifluoromethylnicotinic acid,2-chloro-6-trifluoromethyl pyridine-3-carboxylic acid,2-chloro-6-trifluoromethyl-nicotinic acid,2-chloro-6-trifluoromethyl-3-pyridinecarboxylic acid,3-pyridinecarboxylic acid, 2-chloro-6-trifluoromethyl,pubchem13677,acmc-1c67o,2-chloro-6-trifluoromethyl nicotinicacid PubChem CID: 2778210 IUPAC Name: 2-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid SMILES: C1=CC(=NC(=C1C(=O)O)Cl)C(F)(F)F