accessibility menu, dialog, popup

UserName

OHFFF

CAS RN 2822-41-5

IUPAC Name: 2,3,4-trifluorophenol
Synonyms: 2,3,4-trifluorophenol
Molecular Weight (g/mol): 148.08
Molecular Formula: C6H3F3O
InChi Key: IJGSULQFKYOYEU-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results