accessibility menu, dialog, popup

UserName

CAS RN 2835-68-9

H2NONH2

CAS RN 2835-68-9

IUPAC Name: 4-aminobenzamide
Synonyms: 4-aminobenzamide
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: QIKYZXDTTPVVAC-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)