CAS RN 289-95-2
CAS RN 289-95-2
IUPAC Name:
pyrimidine
Synonyms:
pyrimidine miazine dimer pyrimidine PYR metadiazine M-diazine pyrimidine base pyrimidine dimer pyrimidine, dimer
Molecular Weight (g/mol):
80.09
Molecular Formula:
C4H4N2
InChi Key:
CZPWVGJYEJSRLH-UHFFFAOYSA-N
SMILES:
C1=CN=CN=C1
1
–
2
of
2
results
1
Pyrimidine, 98+%
CAS: 289-95-2 Molecular Formula: C4H4N2 Molecular Weight (g/mol): 80.09 MDL Number: MFCD00006059 InChI Key: CZPWVGJYEJSRLH-UHFFFAOYSA-N Synonym: 1,3-diazine,metadiazine,m-diazine,miazine,1,3-diazabenzene,base,1,3-diazin,pyrimidin,unii-k8cxk5q32l,k8cxk5q32l PubChem CID: 9260 ChEBI: CHEBI:16898 IUPAC Name: pyrimidine SMILES: C1=CN=CN=C1
2
Pyrimidine, 99%
CAS: 289-95-2 Molecular Formula: C4H4N2 Molecular Weight (g/mol): 80.09 MDL Number: MFCD00006059 InChI Key: CZPWVGJYEJSRLH-UHFFFAOYSA-N Synonym: 1,3-diazine,metadiazine,m-diazine,miazine,1,3-diazabenzene,base,1,3-diazin,pyrimidin,unii-k8cxk5q32l,k8cxk5q32l PubChem CID: 9260 ChEBI: CHEBI:16898 IUPAC Name: pyrimidine SMILES: C1=CN=CN=C1