accessibility menu, dialog, popup

UserName

CAS RN 2905-23-9

IUPAC Name: 2-chlorobenzene-1-sulfonyl chloride
Synonyms: 2-chlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 211.06
Molecular Formula: C6H4Cl2O2S
InChi Key: KMVZDSQHLDGKGV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)