CAS RN 2922-45-4
CAS RN 2922-45-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
pyridine-3-sulfonamide, TRC
CAS: 2922-45-4 Molecular Formula: C5H6N2O2S Molecular Weight (g/mol): 158.18 IUPAC Name: pyridine-3-sulfonamide SMILES: NS(=O)(=O)c1cccnc1