accessibility menu, dialog, popup

UserName

KOOFFF

CAS RN 2923-16-2

IUPAC Name: potassium trifluoroacetate
Synonyms: potassium trifluoroacetate
Molecular Weight (g/mol): 152.11
Molecular Formula: C2F3KO2
InChi Key: CUNPJFGIODEJLQ-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results