accessibility menu, dialog, popup

UserName

BrOOO

CAS RN 29310-88-1

IUPAC Name: 3-(2-bromoacetyl)-2H-chromen-2-one
Synonyms: 3-(2-bromoacetyl)chromen-2-one
Molecular Weight (g/mol): 267.08
Molecular Formula: C11H7BrO3
InChi Key: NTYOLVNSXVYRTJ-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC2=CC=CC=C2OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results