accessibility menu, dialog, popup

UserName

H3CCH3ClO

CAS RN 2936-08-5

IUPAC Name: 2-propylpentanoyl chloride
Synonyms: 2-propylpentanoyl chloride
Molecular Weight (g/mol): 162.66
Molecular Formula: C8H15ClO
InChi Key: PITHYUDHKJKJNQ-UHFFFAOYSA-N
SMILES: CCCC(CCC)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)