accessibility menu, dialog, popup

UserName

CAS RN 293762-45-5

NNHNHNNH2H2N

CAS RN 293762-45-5

IUPAC Name: 2-[2-(4-aminophenyl)-1H-1,3-benzodiazol-6-yl]-1H-1,3-benzodiazol-6-amine
Synonyms: 2-[2-(4-aminophenyl)-3H-1,3-benzodiazol-5-yl]-3H-1,3-benzodiazol-5-amine
Molecular Weight (g/mol): 340.39
Molecular Formula: C20H16N6
InChi Key: PAGZCEHLFCJSPV-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NC2=CC=C(N)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results