accessibility menu, dialog, popup

UserName

CAS RN 29529-99-5

H3CNCH3NSHNSNH

CAS RN 29529-99-5

IUPAC Name: 6-(dibutylamino)-1,2,3,4-tetrahydro-1,3,5-triazine-2,4-dithione
Synonyms: 6-(dibutylamino)-1,3-dihydro-1,3,5-triazine-2,4-dithione
Molecular Weight (g/mol): 272.43
Molecular Formula: C11H20N4S2
InChi Key: IXDGHAZCSMVIFX-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C1=NC(=S)NC(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results