accessibility menu, dialog, popup

UserName

NN(E)

CAS RN 298-57-7

IUPAC Name: 1-(diphenylmethyl)-4-[(2E)-3-phenylprop-2-en-1-yl]piperazine
Synonyms: cinnarizine aplexal cinnarizine, (e)-isomer cinnarizine l tartrate cinnarizine L-tartrate (1:1) cinazière stutgeron denapol cisaken dimitronal
Molecular Weight (g/mol): 368.52
Molecular Formula: C26H28N2
InChi Key: DERZBLKQOCDDDZ-JLHYYAGUSA-N
SMILES: C(\C=C\C1=CC=CC=C1)N1CCN(CC1)C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)