CAS RN 29823-18-5
CAS RN 29823-18-5
IUPAC Name:
ethyl 7-bromoheptanoate
Synonyms:
ethyl 7-bromoheptanoate
Molecular Weight (g/mol):
237.14
Molecular Formula:
C9H17BrO2
InChi Key:
OOBFNDGMAGSNKA-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCCCCBr
1
–
3
of
3
results
2
Ethyl 7-bromoheptanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ethyl 7-Bromoheptanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.