CAS RN 299-95-6
IUPAC Name:
bis(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol); sulfuric acid
Synonyms:
bis(isoproterenol); sulfuric acid
Molecular Weight (g/mol):
520.59
Molecular Formula:
C22H36N2O10S
InChi Key:
ZOLBALGTFCCTJF-UHFFFAOYNA-N
SMILES:
OS(O)(=O)=O.CC(C)NCC(O)C1=CC=C(O)C(O)=C1.CC(C)NCC(O)C1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Isoprenaline hemisulfate
CAS: 299-95-6 Molecular Formula: C11H17NO3·1/2H2O4S Molecular Weight (g/mol): 520.59 InChI Key: ZOLBALGTFCCTJF-UHFFFAOYSA-N Synonym: isoprenaline sulphate,isoprenaline sulfate,aleudrin,novodrine,aleudrine sulfate,isoproterenol sulfate,dl-isoprenaline sulfate,dl-isoproterenol sulfate,d,l-isoproterenol sulfate,+--isoprenaline sulfate PubChem CID: 8239 IUPAC Name: 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid SMILES: CC(C)NCC(C1=CC(=C(C=C1)O)O)O.CC(C)NCC(C1=CC(=C(C=C1)O)O)O.OS(=O)(=O)O