accessibility menu, dialog, popup

UserName

HNOOH(E/Z)

CAS RN 29953-71-7

IUPAC Name: 3-(1H-indol-3-yl)prop-2-enoic acid
Synonyms: 3-indoleacrylic acid
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: PLVPPLCLBIEYEA-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results