accessibility menu, dialog, popup

UserName

CAS RN 29976-75-8

H3COO

CAS RN 29976-75-8

IUPAC Name: 6-methyl-2-phenyl-4H-chromen-4-one
Synonyms: 6-methyl-2-phenylchromen-4-one
Molecular Weight (g/mol): 236.27
Molecular Formula: C16H12O2
InChi Key: NOQJBXPAMJLUSS-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC(=CC(=O)C2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results