accessibility menu, dialog, popup

UserName

CAS RN 30007-47-7

ONOBrOO

CAS RN 30007-47-7

IUPAC Name: 5-bromo-5-nitro-1,3-dioxane
Synonyms: 5-bromo-5-nitro-1,3-dioxane bronidox
Molecular Weight (g/mol): 212
Molecular Formula: C4H6BrNO4
InChi Key: XVBRCOKDZVQYAY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1(Br)COCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results