CAS RN 300851-67-6
CAS RN 300851-67-6
IUPAC Name:
2-(4-chlorophenyl)-3-methyl-N-(1,3-thiazol-2-yl)butanamide
Synonyms:
2-(4-chlorophenyl)-3-methyl-N-(1,3-thiazol-2-yl)butanamide
Molecular Weight (g/mol):
294.8
Molecular Formula:
C14H15ClN2OS
InChi Key:
AZYDQCGCBQYFSE-UHFFFAOYNA-N
SMILES:
CC(C)C(C(=O)NC1=NC=CS1)C1=CC=C(Cl)C=C1