accessibility menu, dialog, popup

UserName

CAS RN 30093-99-3

CH3H3CON

CAS RN 30093-99-3

IUPAC Name: 4,4-dimethyl-4,5-dihydro-1,3-oxazole
Synonyms: 4,4-dimethyl-5H-1,3-oxazole
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: KOAMXHRRVFDWRQ-UHFFFAOYSA-N
SMILES: CC1(C)COC=N1
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)