accessibility menu, dialog, popup

UserName

NS

CAS RN 3012-37-1

IUPAC Name: (benzylsulfanyl)carbonitrile
Synonyms: benzyl thiocyanate solvat 14 benzylrhodonid tropeolin
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: ABNDFSOIUFLJAH-UHFFFAOYSA-N
SMILES: N#CSCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results