accessibility menu, dialog, popup

UserName

CAS RN 3014-80-0

IUPAC Name: 2-amino-3-methylbutanamide hydrochloride
Synonyms: 2-amino-3-methylbutanamide hydrochloride
Molecular Weight (g/mol): 152.62
Molecular Formula: C5H13ClN2O
InChi Key: XFCNYSGKNAWXFL-UHFFFAOYNA-N
SMILES: Cl.CC(C)C(N)C(N)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results